missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP3 Antibody (3G3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56582
This item is not returnable.
View return policy
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| FoxP3 | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 2 - 5 ug/mL, Flow Cytometry 100 ng/ 10^6 cells in 50 uL | |
| Q9BZS1 | |
| FOXP3 | |
| Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known, but it has been mapped to the N-terminal portion of the protein (NP_001107849). | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| 3G3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, foxp3 flow, foxp3 flow cytometry, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
| Mouse | |
| 47 kDa | |
| 0.1 mg | |
| Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators | |
| 50943 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction