missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP3 Antibody (3G3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
226.00 € - 521.00 €
Specifications
| Antigen | FoxP3 |
|---|---|
| Clone | 3G3 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 2 - 5 ug/mL, Flow Cytometry 100 ng/ 10^6 cells in 50 uL |
| Applications | Western Blot, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18045784
|
Novus Biologicals
NB100-56582SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18759743
|
Novus Biologicals
NB100-56582 |
0.1 mg |
521.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
| FoxP3 | |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators | |
| PBS with 0.05% Sodium Azide | |
| AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, foxp3 flow, foxp3 flow cytometry, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
| FOXP3 | |
| IgG1 κ | |
| Protein G purified | |
| 47 kDa |
| 3G3 | |
| Western Blot 2 - 5 ug/mL, Flow Cytometry 100 ng/ 10^6 cells in 50 uL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Q9BZS1 | |
| 50943 | |
| Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known, but it has been mapped to the N-terminal portion of the protein (NP_001107849). | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title