missing translation for 'onlineSavingsMsg'
Learn More

FRA10AC1 Antibody, Novus Biologicals™

Product Code. 18156781 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18156781 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18156781 Supplier Novus Biologicals Supplier No. NBP216528

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FRA10AC1 Polyclonal specifically detects FRA10AC1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRA10AC1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.42mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris-Glycine (pH7), 20% Glycerol with 0.01% Thimerosal
Gene Alias C10orf4fragile site 10q23.3, chromosome 10 open reading frame 4, F26C11.1-like, FRA10A, fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate1, protein FRA10AC1, rare folic acid-type fragile site, FRA(10)(q23.3), candidate gene 1
Gene Symbols FRA10AC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human C10orf4. The exact sequence is proprietary.
Molecular Weight of Antigen 38 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118924
Test Specificity Human. Expected to cross react based on sequence identity: Bovine (94%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.