missing translation for 'onlineSavingsMsg'
Learn More

Fructose, LoGiCal

Product Code. 30458141
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458141 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458141 Supplier LoGiCal Supplier No. LGCFOR1562.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Fructose
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-(-)-Fructose
CAS 57-48-7
Analyte or Component Names D-(-)-Fructose
Shipping Condition Room Temperature
Molecular Formula C6H12O6
Certifications/Compliance ISO 9001
Synonym Advantose FS 95, D-(-)-Fructose, D-(-)-Levulose, D-Arabino-2-hexulose, Fructose, Fruit Sugar, Fujifructo L 95, Furucton, Hi-Fructo 970, Krystar, Krystar 300, Levulose, Nevulose, Sugar Fruit;
InChI Formula InChI=1 S/C6H12O6/c7-1-3-4(9)5(10)6(11,2-8)12-3/h3-5,7-11 H,1-2H2/t3-,4-,5+,6-/m1/s1
SMILES O[C@]1(CO)O[C@H](CO)[C@@H](O)[C@@H]1 O
IUPAC Name (2 R,3 S,4 S,5 R)-2,5-bis(hydroxymethyl)tetrahydrofuran-2,3,4-triol
Industry Type Forensics
Molecular Weight (g/mol) 180.16
Quantity 10 mg
Formula Weight 180.06 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.