missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FUBI/MNSF beta Antibody, R&D Systems™

Rat Monoclonal Antibody
209.00 € - 487.00 €
Specifications
| Antigen | FUBI/MNSF beta/FAU |
|---|---|
| Clone | 861504 |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18706429
![]() |
R&D Systems
MAB9036-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18753259
![]() |
R&D Systems
MAB9036 |
100 μg |
487.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FUBI/MNSF beta/FAU Monoclonal specifically detects FUBI/MNSF beta/FAU in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| FUBI/MNSF beta/FAU | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 40S ribosomal protein S30, asr1, FAU, FAU1, FAU-encoded ubiquitin-like protein, FBR-MuSV-associated ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed, Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed(fox derived), FLJ22986, Fub1, Fubi, MNSFbeta, monoclonal nonspecific suppressor factor beta, ribosomal protein S30, RPS30, S30, ubiquitin-like protein fubi and ribosomal protein S30, ubiquitin-like-S30 fusion protein | |
| FAU | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 861504 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| P35544 | |
| 2197 | |
| E.coli-derived recombinant human FUBI/MNSF beta Met1-Gly74 Accession # P35544 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human FUBI in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title