missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fumarylacetoacetate hydrolase Polyclonal antibody specifically detects Fumarylacetoacetate hydrolase in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Fumarylacetoacetate hydrolase |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS |
| Gene Alias | beta-diketonase, EC 3.7.1.2, FAA, fumarylacetoacetase, Fumarylacetoacetate hydrolase, fumarylacetoacetate hydrolase (fumarylacetoacetase) |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-419 of human Fumarylacetoacetate hydrolase (NP_000128.1).,, Sequence:, MSFIPVAEDSDFPIHNLPYGVFSTRGDPRPRIGVAIGDQILDLSIIKHLFTGPVLSKHQDVFNQPTLNSFMGLGQAAWKEARVFLQNLLSVSQARLRDDTELRKCAFISQASATMHLPATIGDYTDFYSSRQHATNVGIMFRDKENALMPNWLHLPVGYHGRASSVVVSGTPIRRPMGQMKPDDSKPPVYGACKLLDMELEMAFFVGPGNRLGEPIPISKAHEHIFGMVLMNDWSARDIQKWEYVPLGPFLGKSFGTTVSPWVVPMDALMPFAVPNPKQDPRPLPYLCHDEPYTFDINLSVNLKGEGMSQAATICKSNFKYMYWTMLQQLTHHSVNGCNLRPGDLLASGTISGPEPENFGSMLELSWKGTKPIDLGNGQTRKFLLDGDEVIITGYCQGDGYRIGFGQCAGKVLPALLPS |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?