missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24687
This item is not returnable.
View return policy
Description
Furin Polyclonal specifically detects Furin in Human, Mouse, Rat, Bovine, Canine, Equine, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Furin | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and xenopus and 95% homologous in cow, mouse, rat, chicken and 89% homologous in dog. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 5 ug/ml | |
| P09958 | |
| FURIN | |
| A portion of amino acids 740-790 of human Furin was used as the immunogen. | |
| 0.1 mg | |
| Golgi Apparatus Markers, Stem Cell Signaling Pathway | |
| 5045 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction