missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 361.00 €
Specifications
| Antigen | Furin |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18128961
|
Novus Biologicals
NBP2-24687SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799763
|
Novus Biologicals
NBP2-24687 |
0.1 mg |
361.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Furin Polyclonal specifically detects Furin in Human, Mouse, Rat, Bovine, Canine, Equine, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Furin | |
| Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Primate | |
| P09958 | |
| 5045 | |
| A portion of amino acids 740-790 of human Furin was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers, Stem Cell Signaling Pathway | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1 | |
| FURIN | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and xenopus and 95% homologous in cow, mouse, rat, chicken and 89% homologous in dog. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title