missing translation for 'onlineSavingsMsg'
Learn More

Furosemide, Mikromol™

Product Code. 18072110
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072110 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18072110 Supplier Mikromol Supplier No. MM0014.000250
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Furosemide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furosemide
CAS 54-31-9
Analyte or Component Names Furosemide
Shipping Condition Room Temperature
Molecular Formula C12 H11 Cl N2 O5 S
Certifications/Compliance ISO 17034
Synonym Benzoic acid, 5-(aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]-, Anthranilic acid, 4-chloro-N-furfuryl-5-sulfamoyl-, 2-Furfurylamino-4-chloro-5-sulfamoylbenzoic acid, 4-Chloro-N-(2-furylmethyl)-5-sulfamoylanthranilic acid, 4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfamyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 5-(Aminosulfonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, Aisemide, Aldic, Aluzine, Anfuramaide, Apo-Frusemide, Apo-Furosemide, Aquamide, Aquarid, Aquasin, Arasemide, Beronald, Bioretic, Cetasix, Desal, Desdemin, Dirine, Disal, Discoid, Disemide, Diural, Diuresal, Diuretic salt, Diurin, Diurolasa, Diusemide, Diusil, Dranex, Dryptal, Durafurid, Errolon, Eutensin, Fluidrol, Franyl, Frumex, Frumide, Frusedan, Frusema, Frusemide, Frusemin, Frusenex, Frusetic, Frusid, Fulsix, Fuluvamide, Furanthril, Furanthryl, Furantril, Furanturil, Furesis, Furetic, Furex, Furix, Furmid, Furo-Basan, Furo-Puren, Furocot, Furodiurol, Furomen, Furomex, Furomide M.D., Furorese, Furosan, Furosedon, Furosemid, Furosemide, Furosix, Furoter, Furovite, Fusid, Golan, Hissuflux, Hydrex, Hydro, Hydro-rapid, Impugan, Katlex, Kofuzon, Kutrix, LB 502, Lasemid, Lasex, Lasiletten, Lasilix, Lasix, Lasix Retard, Laxur, Liside, Lowpston, Lowpstron, Macasirool, Marsemide, Mirfat, NSC 269420, Naclex, Nadis, Nelsix, Nephron, Nicorol, Odemase, Odemex, Oedemex, Prefemin, Profemin, Promedes, Promide, Radisemide, Radonna, Retep, Rosemide, Rusyde, Salinex, Salix, Salurid, Seguril, Selectofur, Transit, Trofurit, Uremide, Urenil, Uresix, Urex, Urex M, Urian, Uridon, Yidoli, Zafurida
InChI Formula InChI=1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)
SMILES NS(=O)(=O)c1cc(C(=O)O)c(NCc2occc2)cc1Cl
IUPAC Name 4-chloro-2-(furan-2-ylmethylamino)-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 330.74
Quantity 250 mg
Formula Weight 330.0077 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.