missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYB/ADAP/SLAP130 Antibody (S03-0H1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19640-100ul
This item is not returnable.
View return policy
Description
FYB/ADAP/SLAP130 Monoclonal antibody specifically detects FYB/ADAP/SLAP130 in Human samples. It is validated for Western Blot
Specifications
| FYB/ADAP/SLAP130 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| S03-0H1 | |
| Unconjugated | |
| ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein | |
| A synthetic peptide of human FYB/ADAP/SLAP130 (Uniprot # O15117) | |
| 100 μg | |
| Immunology | |
| 2533 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction