missing translation for 'onlineSavingsMsg'
Learn More

FYB/ADAP/SLAP130 Antibody (S03-0H1), Novus Biologicals™

Rabbit Monoclonal Antibody

421.00 € - 645.00 €

Specifications

Antigen FYB/ADAP/SLAP130
Clone S03-0H1
Concentration 0.3 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18676483
View Documents
Novus Biologicals
NBP3-19640-100ul
100 μg
645.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18615903
View Documents
Novus Biologicals
NBP3-19640-50ul
50 μg
421.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FYB/ADAP/SLAP130 Monoclonal antibody specifically detects FYB/ADAP/SLAP130 in Human samples. It is validated for Western Blot
Specifications

Specifications

FYB/ADAP/SLAP130
0.3 mg/mL
Monoclonal
Purified
RUO
Human
ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein
A synthetic peptide of human FYB/ADAP/SLAP130 (Uniprot # O15117)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S03-0H1
Western Blot
Unconjugated
Rabbit
Immunology
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
2533
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.