missing translation for 'onlineSavingsMsg'
Learn More

G protein alpha Inhibitor 2 Antibody (S05-7I1), Novus Biologicals™

Product Code. 18695043 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18695043 50 μg 50µL
18658493 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18695043 Supplier Novus Biologicals Supplier No. NBP31986850ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

G protein alpha Inhibitor 2 Monoclonal antibody specifically detects G protein alpha Inhibitor 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen G protein alpha Inhibitor 2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S05-7I1
Concentration 0.3 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin
Formulation 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias alpha-2 subunit, guanine nucleotide binding protein (G protein), alpha inhibiting activitypolypeptide 2
Host Species Rabbit
Immunogen A synthetic peptide of human G protein alpha Inhibitor 2 (Uniprot # P04899)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2771
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.