missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G9a/EHMT2 Antibody (SR1247), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | G9a/EHMT2 |
|---|---|
| Clone | SR1247 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697094
|
Novus Biologicals
NBP3-22088-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18694534
|
Novus Biologicals
NBP3-22088-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G9a/EHMT2 Monoclonal antibody specifically detects G9a/EHMT2 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| G9a/EHMT2 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ankyrin repeat-containing protein, BAT8, C6orf30, chromosome 6 open reading frame 30, DKFZp686H08213, EC 2.1.1.-, EC 2.1.1.43, euchromatic histone-lysine N-methyltransferase 2HLA-B-associated transcript 8, FLJ35547, G9A histone methyltransferase, G9AEm:AF134726.3, H3-K9-HMTase 3, Histone H3-K9 methyltransferase 3, histone-lysine N-methyltransferase, H3 lysine-9 specific 3, HLA-B associated transcript 8, KMT1CNG36/G9a, Lysine N-methyltransferase 1C, NG36, Protein G9a | |
| A synthesized peptide derived from human G9a/EHMT2 (Uniprot #: Q96KQ7) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SR1247 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 10919 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title