missing translation for 'onlineSavingsMsg'
Learn More

GABA-A R rho 2 Antibody, Novus Biologicals™

Product Code. 18251416 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18251416 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18251416 Supplier Novus Biologicals Supplier No. NBP180061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GABA-A R rho 2 Polyclonal specifically detects GABA-A R rho 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABA-A R rho 2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P28476
Gene Alias GABA(A) receptor subunit rho-2, GABA-C receptor, rho-2 subunit, gamma-aminobutyric acid (GABA) receptor, rho 2, gamma-aminobutyric acid receptor subunit rho-2
Gene Symbols GABRR2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human GABRR2 (NP_002034). Peptide sequence SNKSMTFDGRLVKKIWVPDVFFVHSKRSFTHDTTTDNIMLRVFPDGHVLY.
Molecular Weight of Antigen 54 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2570
Test Specificity Expected identity based on immunogen sequence: Canine: 100%; Chicken: 92%; Guinea pig: 92%; Mouse: 92%; Rabbit: 92%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.