missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galactosylceramidase/GALC Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33411-20ul
This item is not returnable.
View return policy
Description
Galactosylceramidase/GALC Monoclonal antibody specifically detects Galactosylceramidase/GALC in Human samples. It is validated for ELISA,Immunocytochemistry/Immunofluorescence
Specifications
| Galactosylceramidase/GALC | |
| Monoclonal | |
| ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| EC 3.2.1.46, galactocerebrosidase, Galactocerebroside beta-galactosidase, Galactosylceramidase, galactosylceramidase (Krabbe disease), Galactosylceramide beta-galactosidase, galactosylceraminidase, GALCERase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human Galactosylceramidase/GALC (NP_000144.2).,, Sequence:, GGAYVLDDSDGLGREFDGIGAVSGGGATSRLLVNYPEPYRSQILDYLFKPNFGASLHILKVEIGGDGQTTDGTEPSHMHYALDENYFRGYEWWLMKEAKKRNPNITLIGLPWSFPGWLGKGFDWPYVNLQLTAYYVVTWIVGAKRYHDLDIDYIGIWNERSYNANYIKILRKMLNYQGLQRVKIIASDNLWESISASMLLDAELFKVVDVIGAHYPGTHSAKDAKLTGKKLWSSEDFSTLNSDMGAGCWGRILNQNYINGY | |
| 20 μL | |
| Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 2581 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction