missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galactosylceramidase/GALC Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
224.00 € - 530.00 €
Specifications
| Antigen | Galactosylceramidase/GALC |
|---|---|
| Dilution | ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233114
|
Novus Biologicals
NBP3-33411-20ul |
20 μL |
224.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231152
|
Novus Biologicals
NBP3-33411-100ul |
100 μL |
530.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Galactosylceramidase/GALC Monoclonal antibody specifically detects Galactosylceramidase/GALC in Human samples. It is validated for ELISA,Immunocytochemistry/ImmunofluorescenceSpecifications
| Galactosylceramidase/GALC | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 2581 | |
| IgG | |
| Affinity purified |
| ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.2.1.46, galactocerebrosidase, Galactocerebroside beta-galactosidase, Galactosylceramidase, galactosylceramidase (Krabbe disease), Galactosylceramide beta-galactosidase, galactosylceraminidase, GALCERase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-300 of human Galactosylceramidase/GALC (NP_000144.2).,, Sequence:, GGAYVLDDSDGLGREFDGIGAVSGGGATSRLLVNYPEPYRSQILDYLFKPNFGASLHILKVEIGGDGQTTDGTEPSHMHYALDENYFRGYEWWLMKEAKKRNPNITLIGLPWSFPGWLGKGFDWPYVNLQLTAYYVVTWIVGAKRYHDLDIDYIGIWNERSYNANYIKILRKMLNYQGLQRVKIIASDNLWESISASMLLDAELFKVVDVIGAHYPGTHSAKDAKLTGKKLWSSEDFSTLNSDMGAGCWGRILNQNYINGY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title