Learn More
Gamma Catenin Mouse, anti-Human, CoraLite Plus 488, Clone: 1A5D10, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | Gamma Catenin |
|---|---|
| Clone | 1A5D10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18036205
|
Proteintech
CL488-66445-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18028085
|
Proteintech
CL488-66445-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Gamma Catenin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Gamma Catenin. This antibody recognizes human, rat, pig, canine antigen. The Gamma Catenin antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| Gamma Catenin | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 3728 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1A5D10 | |
| Western Blot | |
| Liquid | |
| Human, Rat, Pig, Canine | |
| ARVD12, Catenin gamma, CTNNG, Desmoplakin 3, Desmoplakin III, DP3, DPIII, junction plakoglobin, JUP, PDGB, PKGB, γ-Catenin | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.