missing translation for 'onlineSavingsMsg'
Learn More

gamma-Glutamylcyclotransferase/CRF21/GGCT Antibody (04), R&D Systems™

Product Code. 18611167 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18611167 100 μg 100µL
18625916 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18611167 Supplier Novus Biologicals Supplier No. NBP306551100ul
  • ONLINE EXCLUSIVE PRICE
    345.81 €
    307.65 € / 100µL
    Save 38.16 €
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

gamma-Glutamylcyclotransferase/CRF21/GGCT Monoclonal specifically detects gamma-Glutamylcyclotransferase/CRF21/GGCT in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma-Glutamylcyclotransferase/CRF21/GGCT
Applications ELISA
Classification Monoclonal
Clone 04
Conjugate Unconjugated
Dilution ELISA 1:1000-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias gamma-glutamylcyclotransferase, GCTG, GGC, MGC3077
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human gamma-Glutamylcyclotransferase/CRF21/GGCT (Uniprot#: O75223-1; Met1-Leu188).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 79017
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.