missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ganciclovir Di-O-acetate, Mikromol™
Description
Ganciclovir Di-O-acetate
Specifications
Specifications
| Chemical Name or Material | Ganciclovir Di-O-acetate |
| CAS | 86357-19-9 |
| Analyte or Component Names | Ganciclovir Di-O-acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H17 N5 O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 9-[[2-(Acetyloxy)-1-[(acetyloxy)methyl]ethoxy]methyl]-2-amino-1,9-dihydro-6H-purin-6-one, Ganciclovir Di-O-acetate, 6H-Purin-6-one, 9-[[2-(acetyloxy)-1-[(acetyloxy)methyl]ethoxy]methyl]-2-amino-1,9-dihydro- |
| InChI Formula | InChI=1S/C13H17N5O6/c1-7(19)22-3-9(4-23-8(2)20)24-6-18-5-15-10-11(18)16-13(14)17-12(10)21/h5,9H,3-4,6H2,1-2H3,(H3,14,16,17,21) |
| SMILES | CC(=O)OCC(COC(=O)C)OCn1cnc2C(=O)NC(=Nc12)N |
| IUPAC Name | [3-acetyloxy-2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]propyl] acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?