missing translation for 'onlineSavingsMsg'
Learn More

GAPDH Antibody, Novus Biologicals™

Product Code. 18781524 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18781524 0.025 mg 25µg
18750374 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18781524 Supplier Novus Biologicals Supplier No. NB10056875SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 93 publications

GAPDH Polyclonal specifically detects GAPDH in Human, Mouse, Rat, Porcine, Canine, Drosophila, Feline, Hamster, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAPDH
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoblot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Simple Western 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoblotting
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P04406
Gene Alias aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH
Gene Symbols GAPDH
Host Species Rabbit
Immunogen Amino acids 73-87 PITIFQERDPSKIKW of glyceraldehyde 3-phosphate dehydrogenase protein were used as the immunogen of this GAPDH antibody.
Molecular Weight of Antigen 36 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 2597
Test Specificity Cat, Chimpanzee, Drosophila, Human, Macaque, Mouse, Pig and Rat. The amino acid sequence used as immunogen is 100% homologous in human, baboon, chimp and macaque, 93% in pig, rat and gerbil, and 86% mouse, cat and salamander.
Target Species Human, Mouse, Rat, Pig, Drosophila, Feline, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.