missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH (Human Specific) Rabbit anti-Human, CoraLite 647, Clone: 2G21, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech CL647-80570-100UL
This item is not returnable.
View return policy
Description
GAPDH (Human Specific) antibody apecifically detetects GAPDH (Human Specific) antigen in Human Samples. It is validated for Western Blot.Specifications
| GAPDH (Human Specific) | |
| 2G21 | |
| CoraLite 647 | |
| BC004109 | |
| Rabbit | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
| Western Blot | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| GAPDH | |
| GAPDH (Human Specific) fusion protein Ag0766 | |
| 100 μL | |
| 2597 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction