missing translation for 'onlineSavingsMsg'
Learn More

GB/T 21312-2007 14 Quinolones 20 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30322998
Change view
Click to view available options
Quantity:
1.5 mL
Unit Size:
1.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30322998 1.5 mL 1.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322998 Supplier Dr. Ehrenstorfer Supplier No. DREA50000757ME
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cinoxacin∣Ciprofloxacin∣Danofloxacin∣Enoxacin∣Enrofloxacin∣Flumequine∣Lomefloxacin hydrochloride∣Nalidixic acid∣Norfloxacin∣Ofloxacin∣Oxolinic acid∣Pefloxacin∣Pipemidic acid∣Sarafloxacin hydrochloride
CAS 28657-80-9, 85721-33-1, 112398-08-0, 74011-58-8, 93106-60-6, 42835-25-6
Analyte or Component Names Cinoxacin∣Ciprofloxacin∣Danofloxacin∣Enoxacin∣Enrofloxacin∣Flumequine∣Lomefloxacin hydrochloride∣Nalidixic acid∣Norfloxacin∣Ofloxacin∣Oxolinic acid∣Pefloxacin∣Pipemidic acid∣Sarafloxacin hydrochloride
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1.5 mL
Concentration or Composition (by Analyte or Components) Cinoxacin 20 μg/mL∣Ciprofloxacin 20 μg/mL∣Danofloxacin 20 μg/mL∣Enoxacin 20 μg/mL∣Enrofloxacin 20 μg/mL∣Flumequine 20 μg/mL∣Lomefloxacin hydrochloride 20 μg/mL∣Nalidixic acid 20 μg/mL∣Norfloxacin 20 μg/mL∣Ofloxacin 20 μg/mL∣Oxolinic acid 20 μg/mL∣Pefloxacin 20 μg/mL∣Pipemidic acid 20 μg/mL∣Sarafloxacin hydrochloride 20 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.