Learn More
Novus Biologicals™ GEF-H1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30801-50ug
 View more versions of this product
Product Code. 30067199
  • 213.00 € / 50µg
Description
GEF-H1 Polyclonal antibody specifically detects GEF-H1 in Mouse samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GEF-H1
Polyclonal
Western Blot : 4 μg/mL, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
GEF, GEFH1, GEF-H1DKFZp547P1516, Guanine nucleotide exchange factor H1, KIAA0651DKFZp547L106, LFP40FLJ45440, Microtubule-regulated Rho-GEF, P40, Proliferating cell nucleolar antigen p40, rho guanine nucleotide exchange factor 2, Rho/Rac guanine nucleotide exchange factor (GEF) 2, Rho/Rac guanine nucleotide exchange factor 2
Antibody developed using protein corresponding to amino acids 2 to 447 of the GEF-H1 protein fused to GST.
50 μg
Cell Cycle and Replication
9181
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
Phosphate buffered saline
Sheep
Ammonium sulfate precipitation
RUO
Primary
Mouse
Purified