missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GEF-H1 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | GEF-H1 |
|---|---|
| Dilution | Western Blot : 4 μg/mL, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067199
|
Novus Biologicals™
NBP3-30801-50ug |
50 μg |
213.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066909
|
Novus Biologicals™
NBP3-30801-250ug |
250 μg |
508.00 €
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GEF-H1 Polyclonal antibody specifically detects GEF-H1 in Mouse samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| GEF-H1 | |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Sheep | |
| Cell Cycle and Replication | |
| Phosphate buffered saline | |
| 9181 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 4 μg/mL, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| GEF, GEFH1, GEF-H1DKFZp547P1516, Guanine nucleotide exchange factor H1, KIAA0651DKFZp547L106, LFP40FLJ45440, Microtubule-regulated Rho-GEF, P40, Proliferating cell nucleolar antigen p40, rho guanine nucleotide exchange factor 2, Rho/Rac guanine nucleotide exchange factor (GEF) 2, Rho/Rac guanine nucleotide exchange factor 2 | |
| Antibody developed using protein corresponding to amino acids 2 to 447 of the GEF-H1 protein fused to GST. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title