missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-56402SS
This item is not returnable.
View return policy
Description
GFP Polyclonal antibody specifically detects GFP in Nonspecies specific samples. It is validated for Western Blot.
Specifications
| GFP | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Amino acids 204-217 of Green Fluorescent Protein were used as immunogen. | |
| 0.025 mg | |
| Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
| The Green Fluorescent Protein amino acid sequence used as immunogen for this antibody is 100% homologous in YFP (Yellow) and CFP (Cyan). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 0.5-2 ug/ml | |
| P45974 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction