missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
213.00 € - 359.00 €
Specifications
| Antigen | GFP |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18768473
|
Novus Biologicals
NB100-56402SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18778983
|
Novus Biologicals
NB100-56402 |
0.1 mg |
359.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot.Specifications
| GFP | |
| Western Blot 0.5-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| P45974 | |
| Amino acids 204-217 of Green Fluorescent Protein were used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
| PBS with 0.05% Sodium Azide | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| IgG | |
| Protein G purified | |
| The Green Fluorescent Protein amino acid sequence used as immunogen for this antibody is 100% homologous in YFP (Yellow) and CFP (Cyan). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title