missing translation for 'onlineSavingsMsg'
Learn More

Glipizide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30328553
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328553 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328553 Supplier Dr. Ehrenstorfer Supplier No. DREA14025950AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glipizide
CAS 29094-61-9
Analyte or Component Names Glipizide
Shipping Condition Room Temperature
Molecular Formula C21 H27 N5 O4 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym N-[2-[4[[[(Cyclohexylamino)carbonyl]amino]sulfonyl]phenyl]ethyl]-5-methylpyrazinecarboxamide, 1-Cyclohexyl-3-[[p-[2-(5-methylpyrazinecarboxamido)
InChI Formula InChI=1S/C21H27N5O4S/c1-15-13-24-19(14-23-15)20(27)22-12-11-16-7-9-18(10-8-16)31(29,30)26-21(28)25-17-5-3-2-4-6-17/h7-10,13-14,17H,2-6,11-12H2,1H3,(H,22,27)(H2,25,26,28)
SMILES Cc1cnc(cn1)C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3
IUPAC Name N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 445.54
Quantity 1 mL
Formula Weight 445.1784 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.