missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCSH Recombinant Rabbit Monoclonal Antibody (24GB1325)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557625
This item is not returnable.
View return policy
Description
PRKCSH Recombinant Monoclonal Antibody for Western Blot, Flow
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.
Specifications
| PRKCSH | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PRKCSH | |
| 80K-H; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; AGE-R2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; G19P1; GIIB; GLU2B; glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; glucosidase II subunit beta; hepatocystin; PCLD; PKCSH; PLD1; PRKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate 80K-H; protein kinase C substrate, 80 Kda protein; VASAP-60 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5589 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1325 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P14314 | |
| PRKCSH | |
| A synthesized peptide derived from human GLU2B (aa 501-528). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction