missing translation for 'onlineSavingsMsg'
Learn More

Glutamine Synthetase Antibody (CL3004) [Alexa Fluor™ 350], Novus Biologicals™

Product Code. 30523627 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30523627 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30523627 Supplier Novus Biologicals Supplier No. NBP343806AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamine Synthetase
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL3004
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P15104, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia, Neurodegeneration, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2752
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.