missing translation for 'onlineSavingsMsg'
Learn More

Glutathione S-Transferase/GST Antibody (GST13) - VHH, R&D Systems™

Product Code. 18604926 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18604926 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18604926 Supplier Novus Biologicals Supplier No. NBP312166
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Llama Monoclonal Antibody

Glutathione S-Transferase/GST Monoclonal specifically detects Glutathione S-Transferase/GST in E. coli samples. It is validated for Dot Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutathione S-Transferase/GST
Applications Dot Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GST13
Conjugate Unconjugated
Dilution Western Blot 1:200, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200, Dot Blot 1 μg/mL
Formulation 1X PBS, pH 7.42, 20% (v/v) Glycerol
Host Species Llama
Immunogen Glutathione S-Transferase/GST
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.