missing translation for 'onlineSavingsMsg'
Learn More

SSEA-1 Antibody (CL6334), R&D Systems™

Product Code. 18602990 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18602990 25 μL 25µL
18366290 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18602990 Supplier Novus Biologicals Supplier No. NBP27648725ul
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

SSEA-1 Monoclonal specifically detects SSEA-1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSEA-1
Applications Western Blot
Classification Monoclonal
Clone CL6334
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias alpha-(1,3)-fucosyltransferase, CD15, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, ELAM-1 ligand fucosyltransferase, ELFT, FCT3AELAM ligand fucosyltransferase, Fucosyltransferase 4, fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific), Fucosyltransferase IV, fucT-IV, FUC-TIV, FUTIV, Galactoside 3-L-fucosyltransferase, Lewis X, LeX, SSEA-1, stage-specific embryonic antigen 1
Gene Symbols FUT4
Host Species Mouse
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: RKPSGAGWEKEWAEAPQEA
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cellular Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2526.0
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.