missing translation for 'onlineSavingsMsg'
Learn More

Glycine Ethyl Ester Hydrochloride, Mikromol

Product Code. 30461712
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461712 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461712 Supplier Mikromol Supplier No. MM1509.040025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Glycine Ethyl Ester Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycine Ethyl Ester Hydrochloride
CAS 623-33-6
Analyte or Component Names Glycine Ethyl Ester Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C4 H9 N O2 . Cl H
Certifications/Compliance ISO 9001
Synonym Glycine, ethyl ester, hydrochloride (1:1), Glycine, ethyl ester, hydrochloride (6CI,8CI,9CI), (2-Ethoxy-2-oxoethyl)ammonium chloride, 279: PN: US20070105806 PAGE: 26 claimed protein, 27: PN: WO2010042547 PAGE: 47 claimed protein, 281: PN: US20050107325 PAGE: 127 claimed protein, Aminoacetic acid ethyl ester hydrochloride, Ethyl 2-aminoacetate hydrochloride, Ethyl aminoacetate hydrochloride, Ethyl glycinate hydrochloride, Glycine ethyl ester hydrochloride, Glycine ethyl ester hydrochloride salt, Glycine ethyl ester monohydrochloride, Glycine ethyl ether hydrochloride, Glycine hydrochloride ethyl ester, L-Glycine ether ester hydrochloride
InChI Formula InChI=1S/C4H9NO2.ClH/c1-2-7-4(6)3-5;/h2-3,5H2,1H3;1H
Solution Type Neat
SMILES Cl.CCOC(=O)CN
IUPAC Name ethyl 2-aminoacetate;hydrochloride
Molecular Weight (g/mol) 139.58
Quantity 25 mg
Formula Weight 139.04 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.