missing translation for 'onlineSavingsMsg'
Learn More

Glycine Receptor alpha 3 Antibody [FITC], Novus Biologicals™

Product Code. 30570858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30570858 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30570858 Supplier Novus Biologicals Supplier No. NBP346221F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Glycine Receptor alpha 3 Polyclonal antibody specifically detects Glycine Receptor alpha 3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycine Receptor alpha 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias glycine receptor alpha 3 subunit, glycine receptor subunit alpha-3, glycine receptor, alpha 3, glycine receptor, alpha-3 polypeptide, ligand gated ion channel
Host Species Rabbit
Immunogen glycine receptor, alpha 3. (Uniprot# O75311)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8001
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.