missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glymidine Sodium, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Glymidine Sodium |
| CAS | 3459-20-9 |
| Analyte or Component Names | Glymidine Sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 N3 O4 S . Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzenesulfonamide, N-[5-(2-methoxyethoxy)-2-pyrimidinyl]-, sodium salt (1:1), Benzenesulfonamide, N-[5-(2-methoxyethoxy)-2-pyrimidinyl]-, sodium salt (8CI,9CI), Sodium, [N-[5-(2-methoxyethoxy)-2-pyrimidinyl]benzenesulfonamido]- (7CI), BS 717, Glycanol, Glycodiazin, Glycodiazine, Glycodiazine sodium, Glyconormal, Glymidine, Glymidine sodium, Glymidine sodium salt, Gondafon, Lycanol, N-[5-(2-Methoxyethoxy)-2-pyrimidinyl]benzenesulfonamide sodium, Redul, Redul IF, Redul sodium, SH 717, Sodium 2-benzenesulfonylamino-5-(2-methoxyethoxy)pyrimidine, Sodium benzenesulfonamide, N-[5-(2-methoxyethoxy)-2-pyrimidinyl]-, Sodium glycodiazine, Sodium glymidine |
| InChI Formula | InChI=1S/C13H14N3O4S.Na/c1-19-7-8-20-11-9-14-13(15-10-11)16-21(17,18)12-5-3-2-4-6-12;/h2-6,9-10H,7-8H2,1H3;/q-1;+1 |
| Solution Type | Neat |
| SMILES | [Na+].COCCOc1cnc([N-]S(=O)(=O)c2ccccc2)nc1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?