missing translation for 'onlineSavingsMsg'
Learn More

Glypican 1 Antibody (AbAb2-GPC1) [Alexa Fluor™ 700] - Chimeric, Novus Biologicals™

Product Code. 30533066 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30533066 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533066 Supplier Novus Biologicals Supplier No. NBP345126AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glypican 1 Monoclonal antibody specifically detects Glypican 1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 1
Applications ELISA
Classification Monoclonal
Clone AbAb2-GPC1
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1
Host Species Mouse
Immunogen Mice were immunized with human Glypican 1 (Uniprot#: P35052)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2817
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.