missing translation for 'onlineSavingsMsg'
Learn More

Glypican 3 Antibody (GPC3/8363R), Novus Biologicals™

Product Code. 18604525 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18604525 20 μg 20µg
18682915 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18604525 Supplier Novus Biologicals Supplier No. NBP32028520ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Glypican 3 Monoclonal antibody specifically detects Glypican 3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GPC3/8363R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10mM PBS with 0.05% BSA
Gene Alias DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS
Host Species Rabbit
Immunogen Recombinant fragment (around aa400-550) of human Glypican 3 protein (exact sequence is proprietary)
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 2719
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.