missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GlyT2/SLC6A5 Monoclonal antibody specifically detects GlyT2/SLC6A5 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | GlyT2/SLC6A5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S08-1A3 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | GlyT2, GLYT-2, GLYT2GlyT-2, NET1, sodium- and chloride-dependent glycine transporter 2, solute carrier family 6 (neurotransmitter transporter, glycine), member 5, Solute carrier family 6 member 5 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human GlyT2/SLC6A5 (Uniprot # Q9Y345) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?