missing translation for 'onlineSavingsMsg'
Learn More

GM-CSF Antibody Pair (HRP), Novus Biologicals™

Product Code. 18606161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18606161 5 Plates Pack of 5
18211617 15 Plates Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18606161 Supplier Novus Biologicals Supplier No. NBP2788445Plates
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Solid phase sandwich ELISA for the quantitative determination of Human GM-CSF.

GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for ELISA, Sandwich ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GM-CSF
Classification Monoclonal
Conjugate HRP
Dilution Sandwich ELISA
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Gene Symbols CSF2
Host Species Mouse
Quantity 5 Plates
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology
Gene ID (Entrez) 1437
Test Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human GM-CSF.
Content And Storage Storage of components varies. See protocol for specific instructions.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.