missing translation for 'onlineSavingsMsg'
Learn More

GM2 Ganglioside Human anti-Human, Clone: L55-81, R&D Systems™

Product Code. 18614921 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
25 μg
Unit Size:
0.05mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18614921 25 μg 25µg
18263997 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18614921 Supplier Novus Biologicals Supplier No. NBP28127725ug
  • ONLINE EXCLUSIVE PRICE
    521.79 €
    459.90 € / 25µg
    Save 61.89 €
    11% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Human Monoclonal Antibody

GM2 Ganglioside Monoclonal antibody specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay, Cell depletion
TRUSTED_SUSTAINABILITY

Specifications

Antigen GM2 Ganglioside
Applications ELISA, Immunohistochemistry, Immunofluorescence, Functional Assay, Depletion Assay
Classification Monoclonal
Clone L55-81
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Functional, Depletion Assay
Formulation PBS
Gene Alias GM2 (ganglioside), Tay-Sachs ganglioside
Host Species Human
Immunogen L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma.
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Product Type Antibody
Form Purified
Isotype IgM κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.