Learn More
Thermo Scientific™ GMP Startup, Calibration, IQOQ + PT (1 Temp Mapping) and Preventive Maintenance for Cold Storage Unit (New Product)
Product Code. 31703118
Change view
Click to view available options
For Use With (Equipment):
ULT Freezers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31703118 ULT Freezers Each
1 options
Product Code. 31703118 Supplier Thermo Scientific™ Supplier No. GMPCOLDNP
Upgrade Your Lab with Thermo Scientific Equipment
Use promo code 27943
View all promotions
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This GMP service plan supports Cold Storage equipment during the new product stage. Includes priority CM support (NBD in select locations), all-inclusive parts, repair, E-learning, no-charge requalification guarantee, 1 hr. free consultation, dedicated coordinator, ensuring minimized downtime.
GMP Service Plan - New Product (NP) (Part of the Regulatory Assurance Plans)
Overview
  • Tailored for Cold storage equipment at the new equipment stage.
  • Delivers comprehensive startup, qualification, and preventive maintenance support for GMP compliance.
Included Services
  • Start-up: Ensures equipment is installed, connected, and fully operational.
  • Calibration: Confirms accurate performance within specified limits and maintains ISO 9001 compliance.
  • Installation qualification (IQ): Verifies installation site and conditions meet manufacturer requirements. Confirms equipment and accessories arrive undamaged and properly assembled.
  • Operational qualification (OQ): Verifies installed equipment operates as intended according to manufacturer specifications, including key tests such as door ajar, power failure, and temperature alerts.
  • Performance testing (PT): Custom test such as 1 temperature mapping to ensure user specifications are met.
  • Preventive maintenance: Routine inspections to maintain installation-level performance. Includes inspection, cleanup, worn part replacement, performance testing, and tuning.
Additional Benefits
  • Priority enhanced support commitment (Next business day) in select locations
  • All-inclusive repair warranty and coverage, including spare parts.
  • Dedicated account coordinator for personalized support.
  • 1-hour consultation for compliance and service support guidance.
  • Unlimited E-learning access.
  • 50% discount on remote monitoring solutions and optional add-ons (temp map/requalification)
Customer Outcomes
  • Seamless startup with validated documentation.
  • Reliable performance and minimized downtime.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ULT Freezers
Product Type Service
Service Offering Preventative Maintenance