missing translation for 'onlineSavingsMsg'
Learn More

GnRHR Antibody (GNRHR/768), Allophycocyanin, R&D Systems™

Product Code. 18604996 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18604996 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18604996 Supplier Novus Biologicals Supplier No. NBP311436APC
  • ONLINE EXCLUSIVE PRICE
    840.99 €
    731.85 € / 0.1mL
    Save 109.14 €
    12% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

GnRHR Monoclonal specifically detects GnRHR in Human samples. It is validated for Flow Cytometry∣Immunocytochemistry/Immunofluorescence∣Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GnRHR
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GNRHR/768
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor
Host Species Mouse
Immunogen Recombinant full-length human GNRHR protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 2798
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.