missing translation for 'onlineSavingsMsg'
Learn More

GOLGB1/Giantin Antibody, DyLight 488, Novus Biologicals™

Product Code. 18449747 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18449747 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18449747 Supplier Novus Biologicals Supplier No. NBP222322G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GOLGB1/Giantin Polyclonal antibody specifically detects GOLGB1/Giantin in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen GOLGB1/Giantin
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 372 kDa Golgi complex-associated protein, GCP, GCP372GIANTIN, giantin, golgi autoantigen, golgin subfamily b, macrogolgin (with transmembrane signal)1,macrogolgin, golgi integral membrane protein 1, golgin B1, golgin B1, golgi integral membrane protein, Golgin subfamily B member 1, GOLIM1, Macrogolgin
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 950 to 981 of human Golgin B1 using the numbering given in entry NP_004478.3 (GeneID 2804).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2804
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.