missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgi Complex Antibody (SPM581), Novus Biologicals™
Mouse Monoclonal Antibody
265.00 € - 474.00 €
Specifications
| Antigen | Golgi Complex |
|---|---|
| Clone | SPM581 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/mL, Flow Cytometry 0.5-1.0 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18456511
|
Novus Biologicals
NBP2-32853-0.1mg |
0.1 mg |
474.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18478871
|
Novus Biologicals
NBP2-32853-0.02mg |
0.02 mg |
265.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Golgi Complex Monoclonal antibody specifically detects Golgi Complex in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Golgi Complex | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| AI593353;AW537898;CFR;CFR1;CFR-1;ESL1;ESL-1;GLG1;MG160;MG-160;Selel | |
| IgG1 κ | |
| Protein A or G purified |
| SPM581 | |
| Western Blot 1-2 ug/mL, Flow Cytometry 0.5-1.0 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| SU-DHL-1 large cell lymphoma cells. | |
| Primary | |
| Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title