missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GOLIM4 Recombinant Rabbit Monoclonal Antibody (23GB5575), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553900
This item is not returnable.
View return policy
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a type II Golgi-resident protein. It may process proteins synthesized in the rough endoplasmic reticulum and assist in the transport of protein cargo through the Golgi apparatus.
Specifications
| GOLIM4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GOLIM4 | |
| 130 kDa golgi-localized phosphoprotein; 3110027H23Rik; cis Golgi-localized calcium-binding protein; decapacitation factor 10; Df10; GIMPc; Golgi integral membrane protein 4; Golgi integral membrane protein, cis; golgi phosphoprotein 4; Golgi phosphoprotein of 130 kDa; golgi-localized phosphoprotein of 130 kDa; GOLIM4; GOLPH4; GPP130; LOW QUALITY PROTEIN: Golgi integral membrane protein 4; P138; type II Golgi membrane protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27333, 310526, 73124 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5575 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00461, Q5BJK8, Q8BXA1 | |
| GOLIM4 | |
| A synthesized peptide derived from human GOLPH4 (50-100AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction