missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GOLIM4 Recombinant Rabbit Monoclonal Antibody (23GB5575), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | GOLIM4 |
|---|---|
| Clone | 23GB5575 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30087391
|
Invitrogen™
MA553900 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089703
|
Invitrogen™
MA553901 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GOLIM4 Recombinant Monoclonal antibody specifically detects GOLIM4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| GOLIM4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O00461, Q5BJK8, Q8BXA1 | |
| 27333, 310526, 73124 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5575 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 130 kDa golgi-localized phosphoprotein; 3110027H23Rik; cis Golgi-localized calcium-binding protein; decapacitation factor 10; Df10; GIMPc; Golgi integral membrane protein 4; Golgi integral membrane protein, cis; golgi phosphoprotein 4; Golgi phosphoprotein of 130 kDa; golgi-localized phosphoprotein of 130 kDa; GOLIM4; GOLPH4; GPP130; LOW QUALITY PROTEIN: Golgi integral membrane protein 4; P138; type II Golgi membrane protein | |
| A synthesized peptide derived from human GOLPH4 (50-100AA). | |
| Primary | |
| -20°C | |
| GOLIM4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title