missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLPH4 Antibody (SR1126), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22031-50ul
This item is not returnable.
View return policy
Description
GOLPH4 Monoclonal antibody specifically detects GOLPH4 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| GOLPH4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1126 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 | |
| GIMPc, GIMPCcis, golgi integral membrane protein 4, Golgi phosphoprotein 4130 kDa golgi-localized phosphoprotein, Golgi phosphoprotein of 130 kDa, GPP130Golgi-localized phosphoprotein of 130 kDa | |
| A synthesized peptide derived from human GOLPH4 (Uniprot #: O00461) | |
| 50 μg | |
| Golgi Apparatus Markers | |
| 27333 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction