missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLPH4 Antibody (SR1126), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | GOLPH4 |
|---|---|
| Clone | SR1126 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18670194
|
Novus Biologicals
NBP3-22031-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610874
|
Novus Biologicals
NBP3-22031-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GOLPH4 Monoclonal antibody specifically detects GOLPH4 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| GOLPH4 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| GIMPc, GIMPCcis, golgi integral membrane protein 4, Golgi phosphoprotein 4130 kDa golgi-localized phosphoprotein, Golgi phosphoprotein of 130 kDa, GPP130Golgi-localized phosphoprotein of 130 kDa | |
| A synthesized peptide derived from human GOLPH4 (Uniprot #: O00461) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1126 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Golgi Apparatus Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 27333 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title