Learn More
Invitrogen™ GPCR/G-protein complex-stabilizing scFv Recombinant Mouse Monoclonal Antibody (scFv16)
Mouse Recombinant Monoclonal Antibody
Brand:  Invitrogen™ 7039761MG
 View more versions of this product
Product Code. 17194679
  • 2020.00 € / 1mg
Description
The scFv antibody stabilizes GPCR/G-protein complexes for cryo-EM of active structures. It recognizes an interface between G alpha and G beta/gamma subunits, and confers resistance to nucleotide exchange-triggered dissociation of the heterotrimeric G-protein. In addition to members of the Gi/o family, this antibody fragment can also be applied to chimeric variants and stabilize other G-protein subtypes including Gs complexes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPCR/G-protein complex-stabilizing scFv
Recombinant Monoclonal
2.0 mg/mL
25mM HEPES with 10% glycerol and no preservative; pH 7.5
Rhodopsin/Gi1.
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Not Available
scFv16
Unconjugated
Mouse
Affinity chromatography