missing translation for 'onlineSavingsMsg'
Learn More

GPER/GPR30 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

401.00 € - 600.00 €

Specifications

Antigen GPER/GPR30
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18679654
View Documents
Novus Biologicals
NBP3-21238-25ul
25 μL
401.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18621964
View Documents
Novus Biologicals
NBP3-21238-100ul
100 μL
600.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GPER/GPR30 Polyclonal antibody specifically detects GPER/GPR30 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

GPER/GPR30
Immunocytochemistry
Unconjugated
Rabbit
Breast Cancer, Cancer, GPCR, Neuroscience
PBS, pH 7.2, 40% glycerol
2852
IgG
Affinity purified
Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Polyclonal
Purified
RUO
Human
CEPRG-protein coupled receptor 30, Chemoattractant receptor-like 2, chemokine receptor-like 2, CMKRL2MGC99678, constitutively expressed peptide-like receptor, DRY12IL8-related receptor DRY12, FEG-1mER, Flow-induced endothelial G-protein coupled receptor 1, G protein-coupled estrogen receptor 1, G protein-coupled receptor 30, GPCR-BR, GPR30GPCR-Br, G-protein coupled estrogen receptor 1, heptahelix receptor, LERGU, LERGU2, leucine rich protein in GPR30 3'UTR, LyGPR, Lymphocyte-derived G-protein coupled receptor, Membrane estrogen receptor
This antibody was developed against Recombinant Protein corresponding to amino acids: MDVTSQARGVGLEMYPGTAQPAAPNTTSPELNLSHPLLGTALANGTGELSEHQQYVIGLFLS
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.