missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPIHBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-55293SS
This item is not returnable.
View return policy
Description
GPIHBP1 Polyclonal specifically detects GPIHBP1 in Human samples. It is validated for Western Blot.
Specifications
| GPIHBP1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen has 78% homology to mouse. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2 ug/ml | |
| Q8IV16 | |
| GPIHBP1 | |
| A synthetic peptide corresponding to an internal region [within residues 100-180] of human GPIHBP1. [Swiss-Prot# Q8IV16] | |
| 0.025 mL | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 338328 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction